missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)germanium(IV), 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11345567
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mg
2 g
10 g
missing translation for 'unitSize'
10g
2g
500mg
This item is not returnable. View return policy

Product Code. 11345567

missing translation for 'mfr': Thermo Scientific Alfa Aesar 042760.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Preparation of nitrides

Preparation of nitrides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7344-40-3
Molecular Formula C8H24GeN4
Molecular Weight (g/mol) 248.938
MDL Number MFCD01863476
InChI Key JKUUTODNPMRHHZ-UHFFFAOYSA-N
Synonym germane, tetrakis dimethylamino, octamethylgermanetetramine, tetrakis dimethylamino germane, tetrakis dimethylamino germanium, dimethyl tris dimethylamino germyl amine
PubChem CID 16684231
IUPAC Name N-methyl-N-[tris(dimethylamino)germyl]methanamine
SMILES CN(C)[Ge](N(C)C)(N(C)C)N(C)C

Specifications

Melting Point 14°C
Color Colorless
Density 1.07 g/mL
Boiling Point 203°C
Flash Point 34°C (93°F)
Odor Amine-like
Linear Formula Ge[N(CH3)2]4
Quantity 10 g
UN Number UN3129
Sensitivity Moisture sensitive
Formula Weight 248.9
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tetrakis (dimethylamino)germanium (IV)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.