missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)diboron, Thermo Scientific™

Product Code. 17322551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17322551 100 g 100g
17302551 5 g 5g
17312551 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17322551 Supplier Thermo Scientific Acros Supplier No. 468541000
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1630-79-1
Molecular Formula C8H24B2N4
Molecular Weight (g/mol) 197.93
MDL Number MFCD00048012
InChI Key KMCDRSZVZMXKRL-UHFFFAOYSA-N
IUPAC Name dimethyl[1,2,2-tris(dimethylamino)diboran-1-yl]amine
SMILES CN(C)B(B(N(C)C)N(C)C)N(C)C

Specifications

Melting Point -33°C
Density 0.926
Boiling Point 55°C to 57°C (2.5 mmHg)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Refractive Index 1.4680 to 1.4760 (20°C, 589 nm)
Quantity 100 g
Formula Weight 197.93
Physical Form Liquid
Chemical Name or Material Tetrakis(dimethylamino)diboron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.