missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyloxymethyl]methane, Mikromol

Product Code. 30458217
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30458217 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458217 Supplier Mikromol Supplier No. MM3963.060100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tetrakis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyloxymethyl]methane, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Plastic additive 09
CAS 6683-19-8
Analyte or Component Names Plastic additive 09
Shipping Condition Room Temperature
Molecular Formula C73 H108 O12
Certifications/Compliance ISO 9001
Synonym 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzenepropanoic acid 1,1'-[2,2-bis[[3-[3,5-bis(1,1-dimethylethyl)-4-hydroxyphenyl]-1-oxopropoxy]methyl]-1,3-propanediyl] ester, 3,5-Di-tert-butyl-4-hydroxyhydrocinnamic acid neopentanetetrayl ester, 3,5-Di-tert-butyl-4-hydroxyhydrocinnamic acid tetraester with pentaerythritol, A 1010, A 1010 (antioxidant), ADK Stab AO 60, AO 1, AO 1010, AO 60, AT 10, AT 10 (antioxidant), Anox 20, Anox 20AM, Antiox 10, Antioxidant 1010, Arenox A 10, BC 1010, BP 101, Carbyltetramethylene tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Chemnox 1010, Chinox 1010, Cyanox 2110, Dovernox 10, Ethanox 310, Evernox 10, Fenozan 22, Fenozan 23, Fero NF 130, Hostanox O 10, IR 1010, IX 1010, Irganox 101, Irganox 1010, Irganox 1010FF, Irganox 1010FP, Irganox 1040, Irganox 1066, Irganox L 101, Irganox RA 1010, Iruga 1010, JY 1010, KY 1010, KY 7910, Longnox 10, Mark AO 60, NF 130, Naugard 10, Neopentanetetrayl 3,5-di-tert-butyl-4-hydroxyhydrocinnamate, PH 1010, Phenosane 23, Quantox 1010, RA 1010, Ralox 630, Richnox 1010, S 1010, Songnox 1010, Sonox 1010, Stabiace PH 1010, Sumilizer BP 101, TTHP, Tetraalkofen BPE, Tinogard TT, Tominox TT, Tominox TT 50, Ultranox 210, Pentaerythritol 4-hydroxy-3,5-di-tert-butylphenylpropionate, Pentaerythritol tetra[3-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propanoate], Pentaerythritol tetra[β-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propionate], Tetrakis[3-(4-hydroxy-3,5-di-tert-butylphenyl)propionyloxymethyl]methane, Tetrakis[[[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyl]oxy]methyl]methane, Tetrakis[methylene(3,5-di-tert-butyl-4-hydroxyhydrocinnamate)]methane, ...
InChI Formula InChI=1S/C73H108O12/c1-65(2,3)49-33-45(34-50(61(49)78)66(4,5)6)25-29-57(74)82-41-73(42-83-58(75)30-26-46-35-51(67(7,8)9)62(79)52(36-46)68(10,11)12,43-84-59(76)31-27-47-37-53(69(13,14)15)63(80)54(38-47)70(16,17)18)44-85-60(77)32-28-48-39-55(71(19,20)21)64(81)56(40-48)72(22,23)24/h33-40,78-81H,25-32,41-44H2,1-24H3
Solution Type Neat
SMILES CC(C)(C)c1cc(CCC(=O)OCC(COC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)(COC(=O)CCc3cc(c(O)c(c3)C(C)(C)C)C(C)(C)C)COC(=O)CCc4cc(c(O)c(c4)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C
IUPAC Name [3-[3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoyloxy]-2,2-bis[3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoyloxymethyl]propyl] 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
Molecular Weight (g/mol) 1177.63
Quantity 100 mg
Formula Weight 1176.7841 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.