missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(2-hydroxyethyl)ammonium bromide, 99%

Product Code. 10771642 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10771642 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10771642 Supplier Thermo Scientific Acros Supplier No. 420470250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4328-04-5 | C8H20BrNO4 | 274.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

CAS 4328-04-5
Melting Point 191°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C8H20BrNO4
Linear Formula (HOCH2CH2)4NBr
MDL Number MFCD00041988
Quantity 25 g
InChI Key HAKHOKJQPCRWAS-UHFFFAOYSA-M
SMILES [Br-].OCC[N+](CCO)(CCO)CCO
IUPAC Name tetrakis(2-hydroxyethyl)azanium bromide
Molecular Weight (g/mol) 274.15
Formula Weight 274.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(2-hydroxyethyl)ammonium bromide, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.