missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV), 99.7% (metals basis), Thermo Scientific Chemicals

Product Code. 11324887
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV) is used as an intermediate to fabricate capacitor devices on the nano scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63370-90-1
Molecular Formula C44H76HfO8
Molecular Weight (g/mol) 911.574
MDL Number MFCD01863471
InChI Key GPCQYWUYUFMJFR-UHFFFAOYSA-N
Synonym hafnium tetramethylheptanedionate, tetrakis dipivaloylmethanato hafnium
PubChem CID 121233202
SMILES CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.[Hf+4]

Specifications

Melting Point 315°C
Quantity 1 g
Solubility Information Soluble in hexane,toluene,hydrocarbons,alcohols,ketones and esters. Insoluble in water.
Formula Weight 910.5
Percent Purity 99.7%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'