missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV), 99.7% (metals basis), Thermo Scientific Chemicals

Product Code. 11344887
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344887 25 g 25g
11324887 1 g 1g
11334887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11344887 Supplier Thermo Scientific Alfa Aesar Supplier No. 041909.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV) is used as an intermediate to fabricate capacitor devices on the nano scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63370-90-1
Molecular Formula C44H76HfO8
Molecular Weight (g/mol) 911.574
MDL Number MFCD01863471
InChI Key GPCQYWUYUFMJFR-UHFFFAOYSA-N
Synonym hafnium tetramethylheptanedionate, tetrakis dipivaloylmethanato hafnium
PubChem CID 121233202
SMILES CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.[Hf+4]

Specifications

Melting Point 315°C
Quantity 25 g
Solubility Information Soluble in hexane,toluene,hydrocarbons,alcohols,ketones and esters. Insoluble in water.
Formula Weight 910.5
Percent Purity 99.7%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.