missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(1-methylethyl) Methylenediphosphonate, Mikromol™

Product Code. 30049893
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049893 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049893 Supplier Mikromol Supplier No. MM3503.02
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tetrakis(1-methylethyl) Methylenediphosphonate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetraisopropyl methylenediphosphonate
CAS 1660-95-3
Analyte or Component Names Tetraisopropyl methylenediphosphonate
Shipping Condition Room Temperature
Molecular Formula C13 H30 O6 P2
Certifications/Compliance ISO 9001
Synonym Medronic Acid Imp. B (EP), P,P'-Methylenebisphosphonic acid, Phosphonic acid, methylenebis-, tetrakis(1-methylethyl) ester (9CI), Phosphonic acid, methylenedi-, tetraisopropyl ester (7CI,8CI), Methylenediphosphonic acid tetraisopropyl ester, NSC 226577, Tetraisopropyl methylenebisphosphonate, Tetraisopropyl methylenediphosphonate, Tetrakis(O-isopropyl) methylenediphosphonate, [(Diisopropoxyphosphinyl)methyl]diisopropoxyphosphine oxide, Medronic Acid Impurity B
InChI Formula InChI=1S/C13H30O6P2/c1-10(2)16-20(14,17-11(3)4)9-21(15,18-12(5)6)19-13(7)8/h10-13H,9H2,1-8H3
SMILES CC(C)OP(=O)(CP(=O)(OC(C)C)OC(C)C)OC(C)C
IUPAC Name 2-[di(propan-2-yloxy)phosphorylmethyl-propan-2-yloxyphosphoryl]oxypropane
Molecular Weight (g/mol) 344.32
Quantity 100 mg
Formula Weight 344.1518 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.