missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydroxyquinone, MedChemExpress
Description
Tetrahydroxyquinone (Tetrahydroxy-1,4-benzoquinone), a primitive anticataract agent, is a redox active benzoquinone. Tetrahydroxyquinone can take part in a redox cycle with semiquinone radicals, leading to the formation of reactive oxygen species (ROS).
Specifications
Specifications
| Chemical Name or Material | Tetrahydroxyquinone |
| CAS | 319-89-1 |
| Color | Black |
| Purity Grade Notes | Research |
| Molecular Formula | C6H4O6 |
| Quantity | 100 mg |
| Synonym | Tetrahydroxy-1,4-benzoquinone Tetrahydroxybenzoquinone |
| Solubility Information | DMSO : 100 mg/mL (581.09 mM; Need ultrasonic) ∣H2O : 1 mg/mL (5.81 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C1C(O)=C(O)C(C(O)=C1O)=O |
| Molecular Weight (g/mol) | 172.09 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?