missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophene, 98%

Product Code. 11333307
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11333307

Brand: Thermo Scientific Alfa Aesar L13908.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 110-01-0 | C4H8S | 88.17 g/mol

Tetrahydrothiophene is used as pharmaceutical and agrochemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrahydrothiophene is used as pharmaceutical and agrochemical intermediates.

Solubility
Immiscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-01-0
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.17
MDL Number MFCD00005476
InChI Key RAOIDOHSFRTOEL-UHFFFAOYSA-N
Synonym tetrahydrothiophene, thiophene, tetrahydro, thiophane, thiacyclopentane, tetramethylene sulfide, thilane, tetrahydrothiophen, pennodorant 1013, thiofan, thiolan
PubChem CID 1127
ChEBI CHEBI:48458
IUPAC Name thiolane
SMILES C1CCSC1

Specifications

Melting Point -96°C
Density 0.999
Boiling Point 120°C to 121°C
Flash Point 19°C (66°F)
Refractive Index 1.504
Quantity 500 g
UN Number UN2412
Beilstein 102392
Merck Index 14,9218
Solubility Information Immiscible in water.
Formula Weight 88.17
Percent Purity 98%
Chemical Name or Material Tetrahydrothiophene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.