missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophene 1-oxide, 97%

Product Code. p-7023692
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11408117

Brand: Thermo Scientific Alfa Aesar A17502.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1600-44-8 | C4H8OS | 104.167 g/mol

In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.

Solubility
Fully miscible in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1600-44-8
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00005477
InChI Key ISXOBTBCNRIIQO-UHFFFAOYSA-N
Synonym tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide
PubChem CID 1128
IUPAC Name thiolane 1-oxide
SMILES C1CCS(=O)C1

Specifications

Density 1.173
Boiling Point 235°C to 237°C
Flash Point 112°C (233°F)
Refractive Index 1.52
Quantity 10 g
Beilstein 105274
Solubility Information Fully miscible in water.
Formula Weight 104.17
Percent Purity 97%
Chemical Name or Material Tetrahydrothiophene 1-oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.