missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophen-3-one, 97%

Product Code. 11473940
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
250 g
Unit Size:
10g
250g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473940 25 g 25g
11463940 5 g 5g
11474453 10 g 10g
11484453 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11473940 Supplier Thermo Scientific Alfa Aesar Supplier No. A11002.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1003-04-9 | C4H6OS | 102.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-04-9
Molecular Formula C4H6OS
Molecular Weight (g/mol) 102.15
MDL Number MFCD00005412
InChI Key DSXFPRKPFJRPIB-UHFFFAOYSA-N
Synonym tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene
PubChem CID 61252
IUPAC Name thiolan-3-one
SMILES O=C1CCSC1

Specifications

Density 1.192
Boiling Point 174°C to 175°C
Flash Point 77°C (170°F)
Odor Characteristic
Refractive Index 1.529
Quantity 25 g
Beilstein 105201
Formula Weight 102.16
Percent Purity 97%
Chemical Name or Material Tetrahydrothiophen-3-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.