missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-4-ylacetic acid, 97%, Thermo Scientific™

Product Code. 10032354
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10032354 5 g Amber Glass Bottle 5g
10749025 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10032354 Supplier Thermo Scientific Maybridge Supplier No. CC67401DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 85064-61-5
Molecular Formula C7H11O3
Molecular Weight (g/mol) 143.16
MDL Number MFCD01631204
InChI Key PBXYNWPYMVWJAH-UHFFFAOYSA-M
Synonym tetrahydropyranyl-4-acetic acid, tetrahydropyran-4-yl-acetic acid, 2-oxan-4-yl acetic acid, 2-tetrahydro-2h-pyran-4-yl acetic acid, tetrahydropyran-4-ylacetic acid, oxan-4-ylacetic acid, tetrahydro-2h-pyran-4-ylacetic acid, 2h-pyran-4-acetic acid, tetrahydro, 2-4-tetrahydropyranyl acetic acid, tetrahydropyran-4-acetic acid
PubChem CID 2773575
IUPAC Name 2-(oxan-4-yl)acetic acid
SMILES [O-]C(=O)CC1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 144.17
Percent Purity 97%
Chemical Name or Material Tetrahydropyran-4-ylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.