missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-4-sulfonyl chloride, 97%

Product Code. 10097649 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10097649 5 g Glass bottle 5g
10250010 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10097649 Supplier Thermo Scientific Acros Supplier No. 429440050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 338453-21-7 | C5H9ClO3S | 184.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338453-21-7
Molecular Formula C5H9ClO3S
Molecular Weight (g/mol) 184.64
InChI Key QQSBPTQNEOJFBO-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-sulfonyl chloride, tetrahydropyran-4-sulfonyl chloride, tetrahydro-pyran-4-sulfonyl chloride, 2h-pyran-4-sulfonylchloride, tetrahydro, tetrahydropyran-4-sulfonylchloride, tetrahydropyran-4-sulphonyl chloride, 2h-pyran-4-sulfonylchloride,tetrahydro, n-5-bromo-2-methoxybenzyl cyclohexanamine hydrobromide
PubChem CID 22637030
IUPAC Name oxane-4-sulfonyl chloride
SMILES C1COCCC1S(=O)(=O)Cl

Specifications

Melting Point 36.0°C to 40.0°C
Color Brown to Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 184.64
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material Tetrahydropyran-4-sulfonyl chloride
compliance-icons
Product Identifier
  • Tetrahydropyran-4-sulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.