missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10072304
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10072304

Brand: Thermo Scientific Maybridge CC41201DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 51673-83-7
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD07779239
InChI Key MQAYFGXOFCEZRW-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-2-carboxylic acid, tetrahydropyran-2-carboxylic acid, 2h-pyran-2-carboxylic acid, tetrahydro, 2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, pubchem19589, acmc-20m8cx, acmc-20m8cy, 2-tetrahydropyranoic acid, tetrahydro-pyran-carboxylic acid, tetrahydro-pyran-2-carboxylic acid
PubChem CID 10964532
IUPAC Name oxane-2-carboxylic acid
SMILES C1CCOC(C1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 1 g
Formula Weight 130.14
Percent Purity 95%
Chemical Name or Material Tetrahydropyran-2-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.