Learn More
(+/-)-Tetrahydrofurfurylamine, 97%
Description
Tetrahydrofurfurylamine was used to study specific interactions between unlike molecules by calculating the molar excess enthalpies. It can be used to produce N-tetrahydrofurfuryl-phthalimide with phthalic acid anhydride by heating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tetrahydrofurfurylamine was used to study specific interactions between unlike molecules by calculating the molar excess enthalpies. It can be used to produce N-tetrahydrofurfuryl-phthalimide with phthalic acid anhydride by heating.
Solubility
Miscible with water.
Notes
Air sensitive & hygroscopic. Store away from oxidizing agents, water/ moisture. Store under dry inert gas.
Specifications
Specifications
| Density | 0.98 |
| Quantity | 100 g |
| Refractive Index | 1.455 |
| UN Number | UN2943 |
| Beilstein | 102729 |
| Boiling Point | 153°C to 154°C |
| Solubility Information | Miscible with water. |
| Formula Weight | 101.15 |
| Percent Purity | 97% |
| Chemical Name or Material | (+/-)-Tetrahydrofurfurylamine |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.