missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfurylamine, 97%, Thermo Scientific Chemicals

Product Code. 10561981
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4795-29-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00005373
InChI Key YNOGYQAEJGADFJ-UHFFFAOYNA-N
Synonym tetrahydrofurfurylamine, 2-aminomethyltetrahydrofuran, 2-tetrahydrofurfurylamine, tetrahydrofuran-2-yl methanamine, 2-furanmethanamine, tetrahydro, furfurylamine, tetrahydro, 2-aminomethyl tetrahydrofuran, usaf q-2, tetrahydro-2-furanmethanamine, oxolan-2-yl-methanamine
PubChem CID 253298
IUPAC Name oxolan-2-ylmethanamine
SMILES NCC1CCCO1

Specifications

Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 153.0°C to 154.0°C (744.0 mmHg)
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4540 to 1.4560
Assay Percent Range 97%
Specific Gravity 0.98
Viscosity 5 mPa.s (20°C)
Beilstein 18, II, 415
Solubility Information Solubility in water: miscible with water. Other solubilities: soluble in alcohol and ether
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material Tetrahydrofurfurylamine, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofurfurylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'