missing translation for 'onlineSavingsMsg'
Learn More

(±)-Tetrahydrofurfuryl Bromide, TRC

Codice prodotto. 30412897
Cambia vista
Click to view available options
Quantity:
50 g
Dimensione della confezione:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
30412897 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30412897 Fornitore Toronto Research Chemicals N. del fornitore TRCT29357050G
 Restricted Item
Estimated Shipment: 10-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Tetrahydrofurfuryl Bromide
CAS 1192-30-9
Molecular Formula C5H9BrO
Quantity 50 g
Synonym 2-(Bromomethyl)tetrahydrofuran (ACI), (RS)-Tetrahydrofurfuryl bromide, (Tetrahydrofuran-2-yl)methyl bromide, (±)-Tetrahydrofurfuryl bromide, 2-(Bromomethyl)oxolane, NSC 93886, Tetrahydrofurfuryl bromide
InChI Formula InChI=1S/C5H9BrO/c6-4-5-2-1-3-7-5/h5H,1-4H2
SMILES BrCC1OCCC1
IUPAC Name 2-(bromomethyl)tetrahydrofuran
Molecular Weight (g/mol) 165.03
Formula Weight 163.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.