missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl bromide, 95%

Product Code. 11452208
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11452208

Brand: Thermo Scientific Alfa Aesar A15192.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1192-30-9 | C5H9BrO | 165.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-30-9
Molecular Formula C5H9BrO
Molecular Weight (g/mol) 165.03
MDL Number MFCD00016894
InChI Key VOHILFSOWRNVJJ-UHFFFAOYNA-N
Synonym tetrahydrofurfuryl bromide, 2-bromomethyl tetrahydrofuran, 2-bromomethyl oxolane, furan, 2-bromomethyl tetrahydro, 2-bromomethyl-tetrahydrofuran, furan, tetrahydro-2-bromomethyl, tetrahydrofurfurylbromide, tetrahydrofrufryl bromide, 2-bromomethyltetrahydrofuran
PubChem CID 14501
IUPAC Name 2-(bromomethyl)oxolane
SMILES BrCC1CCCO1

Specifications

Density 1.452
Boiling Point 169°C to 171°C
Quantity 50 g
Refractive Index 1.488
Beilstein 102717
Formula Weight 165.04
Percent Purity 95%
Chemical Name or Material Tetrahydrofurfuryl bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.