missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofurfuryl Alcohol, TRC
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Specifications
Specifications
| Chemical Name or Material | Tetrahydrofurfuryl alcohol |
| CAS | 97-99-4 |
| Molecular Formula | C5 H10 O2 |
| Quantity | 5 mL |
| Synonym | Furfuryl alcohol','tetrahydro- (8CI), Tetrahydro-2-furanmethanol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydrofurfuryl alcohol, 2-(Hydroxymethyl)tetrahydrofuran, NSC 15434, QO THFA, Rac-(Tetrahydrofuran-2-yl)methanol, THFA, Tetrahydro-2-furancarbinol, Tetrahydro-2-furanylmethanol, Tetrahydro-2-furylmethanol |
| InChI Formula | InChI=1S/C5H10O2/c6-4-5-2-1-3-7-5/h5-6H,1-4H2 |
| SMILES | OCC1CCCO1 |
| IUPAC Name | oxolan-2-ylmethanol |
| Molecular Weight (g/mol) | 102.1317 |
| Formula Weight | 102.0681 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction