missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Honeywell Riedel-de Haën™

Product Code. 15915540 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
45 L
Packaging:
Returnable Stainless Steel Drum
Unit Size:
45 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15915540 45 L Returnable Stainless Steel Drum 45 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15915540 Supplier Honeywell-Riedel-de Haen Supplier No. 7218745LRC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.

TRUSTED_SUSTAINABILITY

Specifications

pH neutral
Melting Point -108°C
CAS 109-99-9
Color Colorless
Quantity 45 L
Packaging Returnable Stainless Steel Drum
Molecular Formula C4H8O
MDL Number MFCD00005356
Boiling Point 65°C (1013 hPa)
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.11
Chemical Name or Material Tetrahydrofuran
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.