missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Extra Pure, SLR, Stabilised with 0.025% BHT, Fisher Chemical™

Product Code. 10040380 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal can
Plastic-coated Glass Bottle
Unit Size:
2.5 L
25 L
5 L
500mL
This item is not returnable. View return policy

Product Code. 10040380

Brand: Fisher Chemical T/0700/PB17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C4H8O, CAS Number-109-99-9, oxacyclopentane, tetraidrofurano, 1,4-epoxybutane, furan, tetrahydro, butylene oxide, hydrofuran, tetrahydrofuranne, tetrahydrofuran, furanidine, tetramethylene oxide, 2.5L, 66 deg.C, CHEBI:26911, Colorless, 72.11g/mol, WYURNTSHIVDZCO-UHFFFAOYSA-N, oxolane, -108.4 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

pH 7 to 8
Melting Point -108.4°C
Color Colorless
Boiling Point 66°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 200mbar at 20°C
Viscosity 0.55cp at 20°C
Formula Weight 72.11g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.