missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-d8 (Reagent grade), CDN

Product Code. 30321008
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Product Code. Quantity unitSize
30321008 10 g 10g
30319716 5 g 5g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30321008

Brand: CDN CDND022610G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrahydrofuran D8
CAS 1693-74-9
Molecular Formula C4 2H8 O
Quantity 10 g
Synonym Furan-d4, tetrahydro-d4- (6 CI,7 CI,8 CI,9 CI), Tetrahydro-d4furan-2,3,4,5-d4, Perdeuterotetrahydrofuran, THF-d8, Tetrahydrofuran-d8, d8-THF
InChI Formula InChI=1 S/C4H8O/c1-2-4-5-3-1/h1-4H2/i1D2,2D2,3D2,4D2
SMILES [2 H]C1([2 H])OC([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H]
IUPAC Name 2,2,3,3,4,4,5,5-octadeuteriooxolane
Molecular Weight (g/mol) 80.16
Formula Weight 80.1077 g/mol
Percent Purity 99 atom % D, min 99% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.