missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran Anhydrous unstabilised, Honeywell™ Riedel de-Haen

Product Code. 15887361
Change view
Click to view available options
Quantity:
100 mL
2.5 L
Unit Size:
100mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15887361 100 mL 100mL
15807371 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15887361 Supplier Honeywell Riedel-de Haën Supplier No. 66205100ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Contain less that 50 ppm water. Honeywell™ Riedel de-Haen Anhydrous Solvents are packaged in specially designed septum-sealed bottles to maintain low water content and ensure product integrity.

  • These solvents are specially produced to a water specification of less than 50 ppm (<0.0050%).
  • Many solvents go well beyond that and are routinely produced to lower than 5 ppm (<0.0005%).
  • All packaging steps and materials are especially designed to retain solvent dryness and ensure product integrity. Before filling, amber glass bottles are oven-dried to remove all traces of water, then filled under dry nitrogen and sealed with a special cap.
  • The water content itself is strictly controlled by coulometric Karl Fischer analysis (Hydranal™) and actual results are printed on the label.
  • This line is packaged in specially designed septum-sealed bottles to maintain low water content and ensure product integrity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Density 0.89 g/cm3 (20°C)
Quantity 100 mL
Flash Point −21°C
Packaging Septum-sealed Bottle
UN Number 2056
Formula Weight 72.11
Percent Purity >99.9%
Chemical Name or Material Tetrahydrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.