missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, ACS Reagent, contains 250 ppm BHT as inhibitor, ≥99.0%, Honeywell Riedel-de Haën™

Product Code. 15666690
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15666690 1 L Glass Bottle 1 L
15608450 4 L Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15666690 Supplier Honeywell-Riedel-de Haen Supplier No. 6767641L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, contains 250 ppm BHT as inhibitor,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C (literature)
CAS Max % <100.0000%
Density 0.889 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point −21°C
Packaging Glass Bottle
Linear Formula C4H8O
UN Number UN2056
Beilstein 102391
Boiling Point 65°C
Formula Weight 72.11
Percent Purity >99%
Grade ACS
Chemical Name or Material Tetrahydrofuran, ACS Reagent, contains 250 ppm BHT as inhibitor, ≥99.0%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.