missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Puriss. p.a., contains 250 ppm BHT as inhibitor, ACS Reagent, Reag. Ph. Eur., ≥99.9%, Solstice

Product Code. 15627370
Change view
Click to view available options
Quantity:
5 L
25 L
4 x 2.5 L
6 x 1 L
175 kg
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
N/A
Stainless Steel Can
Unit Size:
175kg
25 L
5 L
Pack of 4
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15627370 6 x 1 L Glass Bottle Pack of 6
15617370 5 L Aluminum Bottle 5 L
15607370 4 x 2.5 L N/A Pack of 4
15697360 25 L Stainless Steel Can 25 L
15667360 175 kg Metal Drum 175kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15627370 Supplier Honeywell-Riedel-de Haen Supplier No. 873686X1L
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Puriss. p.a., contains 250 ppm BHT as inhibitor, ACS Reagent, Reag. Ph. Eur.,≥99.9%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C
Color Colorless
Density 0.887g/mL at 20°C, 0.889g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.118
Flash Point −17°C (1.4°F)
Ignition Point 610°F
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 114mmHg (15°C), 143mmHg (20°C)
UN Number UN2056
Beilstein 102391
Boiling Point 65°C (1013 hPa)
Formula Weight 72.11g/mol
Percent Purity ≥99.9%
Chemical Name or Material Tetrahydrofuran
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran (stab. with BHT)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.