missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran ≥ 99.8% BAKER ANALYZED™, UHPLC, J.T. Baker™

Product Code. 10354411 Shop All J.T.Baker Products
Change view
Click to view available options
:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10354411 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10354411 Supplier JT Baker Supplier No. 9441.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For HPLC and spectrophotometry

  • Ultraviolet Absorbance (1.00-cm path vs water):
    • at 230 nm max. 1.0
    • at 260 nm max. 0.15
    • at 270 nm max. 0.08
    • at 310 nm max. 0.02

Specifications

Melting Point -108.5°C
CAS 109-99-9
Density 0.888 g/cm³ (20°C)
Quantity 1 L
Flash Point -21.5°C
Residue after Evaporation max. 5 ppm
Assay Percent Range min. 99.8%
Molecular Formula C4H8O
MDL Number MFCD00005356
Water max. 0.02%
Boiling Point 66°C (1013 hPa)
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.11
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.