missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-3-sulfonyl chloride, Thermo Scientific™

Product Code. 10210415
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10210415 5 g Amber Glass Bottle 5g
10311825 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10210415 Supplier Thermo Scientific Maybridge Supplier No. CC76003DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1207346-29-9
Molecular Formula C4H7ClO3S
Molecular Weight (g/mol) 170.607
InChI Key MVZIUOJNVVIOEY-UHFFFAOYSA-N
Synonym tetrahydrofuran-3-sulfonyl chloride, chlorooxolan-3-ylsulfone
PubChem CID 49762850
IUPAC Name oxolane-3-sulfonyl chloride
SMILES C1COCC1S(=O)(=O)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 170.61
Percent Purity 97%
Chemical Name or Material Tetrahydrofuran-3-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.