missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrocannabinol Antibody (094-14066 (THC-003)), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62946IR
This item is not returnable.
View return policy
Description
Tetrahydrocannabinol Monoclonal specifically detects Tetrahydrocannabinol in Non-species specific samples. It is validated for ELISA.
Specifications
| Tetrahydrocannabinol | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| 11-Nor-delta 9-THC-acid conjugated at the carboxyl group. | |
| 0.1 mL | |
| Primary | |
| Non-species specific | |
| Purified |
| ELISA | |
| 094-14066 (THC-003) | |
| ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| NB100-62946 reacts with Delta-9-tetrahydrocannabinol, the main psychoactive substance present in the Cannabis plant. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction