missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydroberberine, MedChemExpress

Product Code. 16499198
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499198 10 mg 10mg
16491608 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499198 Supplier Medchem Express Supplier No. HYN0925
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tetrahydroberberine is an isoquinoline alkaloid isolated from corydalis tuber; has micromolar affinity for dopamine D(2) (pK(i) = 6.08) and 5-HT(1A) (pK(i) = 5.38) receptors but moderate to no affinity for other relevant serotonin receptors (5-HT(1B), 5-HT(1D), 5-HT(3), and 5-HT(4); pK(i) < 5.00).

Tetrahydroberberine is an isoquinoline alkaloid isolated from corydalis tuber

Specifications

Chemical Name or Material Tetrahydroberberine
CAS 522-97-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H21NO4
Quantity 10 mg
Synonym Canadine
Solubility Information DMSO : 25 mg/mL (73.66 mM; Need ultrasonic)
SMILES COC1=CC=C(CC23)C(CN2CCC(C3=C4)=CC5=C4OCO5)=C1OC
Molecular Weight (g/mol) 339.39
Formula Weight 339.39
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.