missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-4-pyrone, TRC

Product Code. 30423712
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423712 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423712 Supplier Toronto Research Chemicals Supplier No. TRCT295465500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrahydro-4H-pyran-4-one
CAS 29943-42-8
Molecular Formula C5 H8 O2
Quantity 500 mg
Synonym 4H-Pyran-4-one','tetrahydro-, Tetrahydro-4H-pyran-4-one, 2,3,5,6-Tetrahydro-4-pyranone, 2,3,5,6-Tetrahydropyran-4-one, 3,4,5,6-Tetrahydro-4H-pyran-4-one, 3,5,6-Trihydro-2H-pyran-4-one, 4-Oxacyclohexanone, 4-Oxotetrahydropyran, 4-Tetrahydropyranone, Dihydro
InChI Formula InChI=1S/C5H8O2/c6-5-1-3-7-4-2-5/h1-4H2
SMILES O=C1CCOCC1
IUPAC Name oxan-4-one
Molecular Weight (g/mol) 100.116
Formula Weight 100.052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.