missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrazolidinone hydrochloride, 98%

Product Code. 10307431 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307431 5 g Glass bottle 5g
10397421 1 g Glass bottle 1g
11976211 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307431 Supplier Thermo Scientific Acros Supplier No. 335490050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1752-88-1 | C3H7ClN2O | 122.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1752-88-1
Molecular Formula C3H7ClN2O
Molecular Weight (g/mol) 122.55
MDL Number MFCD00043787
InChI Key HAYWTXQRYXYRAF-UHFFFAOYSA-N
Synonym 3-pyrazolidinone hydrochloride, pyrazolidin-3-one hydrochloride, 4,5-dihydro-1h-pyrazol-3-ol hydrochloride, 3-pyrazolidinone, monohydrochloride, tetrahydro-3h-pyrazol-3-one hydrochloride, 3-pyrazolidinone hcl, 3-pyrazolidone hydrochloride, ksc493k0l, pyrazolidin-3-on-hydrochloride, pyrazolidin-3-one hydrogen chloride
PubChem CID 2733712
SMILES [H+].[Cl-].O=C1CCNN1

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
IUPAC Name pyrazolidin-3-one;hydrochloride
Formula Weight 122.55
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Pyrazolidinone hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.