missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2-furoic acid, 98+%

Product Code. 10162961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Product Code. Quantity Packaging unitSize
10162961 500 g Glass bottle 500g
10433835 25 g Glass bottle 25g
10637231 100 g Glass bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 10162961

Brand: Thermo Scientific Acros 263145000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16874-33-2 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16874-33-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00022473
InChI Key UJJLJRQIPMGXEZ-UHFFFAOYSA-N
Synonym 2-tetrahydrofuroic acid, tetrahydrofuran-2-carboxylic acid, tetrahydro-2-furoic acid, tetrahydro-2-furancarboxylic acid, 2-furancarboxylic acid, tetrahydro, furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, thfc, tetrahydrofuroic acid, pubchem7088
PubChem CID 86079
IUPAC Name oxolane-2-carboxylic acid
SMILES C1CC(OC1)C(=O)O

Specifications

CAS Min % 99.0
Color Yellow to Brown
CAS Max % 100.0
Density 1.1700g/mL
Boiling Point 128.0°C to 129.0°C (13.0 mmHg)
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4590 to 1.4610
Assay Percent Range 98% min. (GC)
Specific Gravity 1.17
Solubility Information Solubility in water: soluble
Formula Weight 116.12
Percent Purity 98+%
Chemical Name or Material Tetrahydro-2-furoic acid, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.