missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluoroboric Acid Solution, Solstice

Product Code. 15671800 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
500 g
6 x 500 g
Unit Size:
25g
500g
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15671800 500 g 500g
15661800 25 g 25g
15681800 6 x 500 g Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15671800 Supplier Honeywell-Fluka Supplier No. 207934500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

48 wt. % in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16872-11-0
Molecular Formula BF4H
Molecular Weight (g/mol) 87.812
MDL Number MFCD00011345
InChI Key ODGCEQLVLXJUCC-UHFFFAOYSA-O
Synonym fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq
PubChem CID 28118
ChEBI CHEBI:38902
IUPAC Name hydron;tetrafluoroborate
SMILES [H+].[B-](F)(F)(F)F

Specifications

Concentration 48wt. % in H2O
Density 1.4g/mL at 25°C
Vapor Pressure 5mmHg (20°C)
Quantity 500 g
Linear Formula HBF4
UN Number 1775
Formula Weight 87.81g/mol
Chemical Name or Material Tetrafluoroboric Acid Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.