missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyltin, 98%

Product Code. 11394829
Change view
Click to view available options
Quantity:
1 g
2 g
10 g
50 g
Unit Size:
10g
1g
2g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394829 10 g 10g
11304839 1 g 1g
11314839 2 g 2g
11324839 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11394829 Supplier Thermo Scientific Alfa Aesar Supplier No. 071144.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 597-64-8 | C8H20Sn | 234.96 g/mol

Tetraethyltin is used in the electronics industry. Used as catalyst for olefin polymerization and to produce other organotin compounds. Also used as a preservative for wood, textiles, paper, and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-64-8
Molecular Formula C8H20Sn
Molecular Weight (g/mol) 234.96
MDL Number MFCD00009020
InChI Key RWWNQEOPUOCKGR-UHFFFAOYSA-N
Synonym tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa
PubChem CID 11704
IUPAC Name tetraethylstannane
SMILES CC[Sn](CC)(CC)CC

Specifications

Melting Point -112°C
Density 1.187 g/mL
Boiling Point 181°C
Flash Point 53°C (128°F)
Odor Characteristic
Refractive Index 1.473
Quantity 10 g
UN Number UN3384
Solubility Information Insoluble in water; Soluble in alcohol and ether
Formula Weight 234.94
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetraethyltin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.