missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyltin, 97%, ACROS Organics™

Product Code. 10451061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10451061 5mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10451061 Supplier Thermo Scientific Acros Supplier No. 212070050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 597-64-8
Melting Point -112.0°C
Color Colorless
Density 1.1800g/mL
Boiling Point 181.0°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H20Sn
Linear Formula (C2H5)4Sn
Refractive Index 1.4720 to 1.4740
MDL Number MFCD00009020
Quantity 5 mL
Specific Gravity 1.18
Synonym tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa
Solubility Information Solubility in water: insoluble.
InChI Key RWWNQEOPUOCKGR-UHFFFAOYSA-N
SMILES CC[Sn](CC)(CC)CC
IUPAC Name tetraethylstannane
Molecular Weight (g/mol) 234.96
PubChem CID 11704
Formula Weight 234.94
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tetraethyltin
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetraethyltin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.