missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylthiourea, TRC

Product Code. 30427688
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427688 50 mg 50mg
30414410 100 mg -
30410563 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427688 Supplier Toronto Research Chemicals Supplier No. TRCT29285050MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4274-15-1
Molecular Formula C9H20N2S
Molecular Weight (g/mol) 188.33
Synonym 1,1,3,3-tetraethyl-2-thio-urea
IUPAC Name 1,1,3,3-tetraethylthiourea
SMILES CCN(CC)C(=S)N(CC)CC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H20N2S/c1-5-10(6-2)9(12)11(7-3)8-4/h5-8H2,1-4H3
Formula Weight 188.1347
Chemical Name or Material Tetraethylthiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.