missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylsilane, 97%, Thermo Scientific™

Product Code. 15730859 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15730859 5 g 5g
15540534 25 g 25g
15550534 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15730859 Supplier Thermo Scientific Acros Supplier No. 313700050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 631-36-7
Molecular Formula C8H20Si
Molecular Weight (g/mol) 144.34
InChI Key VCZQFJFZMMALHB-UHFFFAOYSA-N
Synonym silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si
PubChem CID 12426
IUPAC Name tetraethylsilane
SMILES CC[Si](CC)(CC)CC

Specifications

Melting Point -82.5°C
Density 0.7600g/mL
Boiling Point 153.0°C to 154.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Si(C2H5)4
Refractive Index 1.4250 to 1.4280
Quantity 5 g
Specific Gravity 0.76
Solubility Information Solubility in water: insoluble.
Formula Weight 144.34
Percent Purity 97%
Chemical Name or Material Tetraethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.