missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylsilane, 97%

Product Code. 11322106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322106 5 g 5g
11332106 25 g 25g
11342106 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11322106 Supplier Thermo Scientific Alfa Aesar Supplier No. L03328.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 631-36-7 | C8H20Si | 144.333 g/mol

Used as the intermediates of OLED, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as the intermediates of OLED, pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-36-7
Molecular Formula C8H20Si
Molecular Weight (g/mol) 144.333
MDL Number MFCD00009019
InChI Key VCZQFJFZMMALHB-UHFFFAOYSA-N
Synonym silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si
PubChem CID 12426
IUPAC Name tetraethylsilane
SMILES CC[Si](CC)(CC)CC

Specifications

Boiling Point 153°C to 155°C
Melting Point -82°C
Quantity 5 g
UN Number UN1993
Beilstein 1732768
Flash Point 26°C (78°F)
Formula Weight 144.33
Percent Purity 97%
Odor Recognizable
Chemical Name or Material Tetraethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.