missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylenepentamine, technical

Product Code. 10456960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10456960 500 g Glass bottle 500g
10205050 250 g Glass bottle 250g
10005773 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10456960 Supplier Thermo Scientific Acros Supplier No. 138115000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-57-2 | C8H23N5 | 189.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-57-2
Molecular Formula C8H23N5
Molecular Weight (g/mol) 189.31
MDL Number MFCD00008168
InChI Key FAGUFWYHJQFNRV-UHFFFAOYSA-N
Synonym tetraethylenepentamine, tetren, 1,4,7,10,13-pentaazatridecane, tetraethylene pentamine, tetraethylpentylamine, 1,11-diamino-3,6,9-triazaundecane, 3,6,9-triazaundecamethylenediamine, 3,6,9-triazaundecane-1,11-diamine, deh 26, unii-yzd1c9kq28
PubChem CID 8197
ChEBI CHEBI:49798
IUPAC Name N'-[2-[2-(2-aminoethylamino)ethylamino]ethyl]ethane-1,2-diamine
SMILES NCCNCCNCCNCCN

Specifications

Melting Point -40°C
Density 0.9900g/mL
Boiling Point 340°C
Flash Point 139°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula HN(CH2CH2NHCH2CH2NH2)2
Refractive Index 1.5055
Quantity 500 g
Specific Gravity 0.99
Solubility Information Solubility in water: soluble
Formula Weight 189.3
Percent Purity ≥95.0%
Grade Technical
Physical Form Liquid
Chemical Name or Material Tetraethylenepentamine, tech.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetraethylenepentamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tetraethylenepentamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.