missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyleneglycol monomethyl ether, 98%

Product Code. 11498876
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498876 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11498876 Supplier Thermo Scientific Alfa Aesar Supplier No. A17021.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23783-42-8 | C9H20O5 | 208.25 g/mol

A very small fraction (less than 5%) may be used for other purposes, such as asphalt anti-stripping applications or as a chemical intermediate in other industrial processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A very small fraction (less than 5%) may be used for other purposes, such as asphalt anti-stripping applications or as a chemical intermediate in other industrial processes.

Solubility
Water solubility 999,999c.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Specifications

CAS 23783-42-8
Density 1.07
Boiling Point 158°C to 160°C (5 mmHg)
Molecular Formula C9H20O5
Refractive Index 1.445
MDL Number MFCD00041756
Quantity 100 g
Synonym 2,5,8,11-tetraoxatridecan-13-ol, tetraethylene glycol monomethyl ether, tetraethyleneglycol monomethyl ether, 3,6,9,12-tetraoxatridecan-1-ol, methyl tetraglycol
Solubility Information Water solubility 999,999 c.
InChI Key ZNYRFEPBTVGZDN-UHFFFAOYSA-N
SMILES COCCOCCOCCOCCO
IUPAC Name 2,5,8,11-tetraoxatridecan-13-ol
Molecular Weight (g/mol) 208.25
PubChem CID 90263
Formula Weight 208.25
Percent Purity 98%
Chemical Name or Material Tetraethyleneglycol monomethyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.