missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene glycol dimethyl ether, 98+%, Thermo Scientific Chemicals

Catalog Number 11377947
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
143-24-8
C10H22O5
222.28
MFCD00008505
ZUHZGEOKBKGPSW-UHFFFAOYSA-N
tetraglyme, tetraethylene glycol dimethyl ether, 2,5,8,11,14-pentaoxapentadecane, dimethoxytetraglycol, glyme 5, ansul ether 181at, dimethoxytetraethylene glycol, nissan uniox mm 200, bis 2-2-methoxyethoxy ethyl ether, methyltetraglyme200
8925
CHEBI:46785
1-methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
COCCOCCOCCOCCOC
This item is not returnable. View return policy
143-24-8
C10H22O5
222.28
MFCD00008505
ZUHZGEOKBKGPSW-UHFFFAOYSA-N
tetraglyme, tetraethylene glycol dimethyl ether, 2,5,8,11,14-pentaoxapentadecane, dimethoxytetraglycol, glyme 5, ansul ether 181at, dimethoxytetraethylene glycol, nissan uniox mm 200, bis 2-2-methoxyethoxy ethyl ether, methyltetraglyme200
8925
CHEBI:46785
1-methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
COCCOCCOCCOCCOC

Tetraethylene glycol dimethyl ether is used in lithium-ion battery technology. It is associated with trifluoroethanol and used as a working pair for organic absorption heat pumps. It acts as a solvent for cleaning and degreasing due to its chemical and thermal stability. Moreover, it is suitable solvent for high temperature reactions due its high boiling point. Further, it is used in the selective adsorption of proteins during the promotion of cell adhesion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -30°C
Density 1.01 g/mL
Boiling Point 275°C to 276°C
Flash Point 140°C (284°F)
Refractive Index 1.433
Quantity 250 g
Merck Index 14,9208
Solubility Information Miscible with water.
Formula Weight 222.28
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Tetraethylene glycol dimethyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'