missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene glycol, 99.5%

Product Code. 10183331 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183331 2.5 L Glass Bottle 2.5 L
10457730 1 L Glass Bottle 1 L
11994731 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183331 Supplier Thermo Scientific Acros Supplier No. 149590025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-60-7 | C8H18O5 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-60-7
Molecular Formula C8H18O5
Molecular Weight (g/mol) 194.23
InChI Key UWHCKJMYHZGTIT-UHFFFAOYSA-N
Synonym tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843
PubChem CID 8200
ChEBI CHEBI:44920
IUPAC Name 2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCO)O

Specifications

Melting Point -4.1°C
Color Yellow
Density 1.1200g/mL
Boiling Point 324.0°C to 330.0°C
Flash Point 176°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass Bottle
Linear Formula O(CH2CH2OCH2CH2OH)2
Refractive Index 1.4580 to 1.4600
Quantity 2.5 L
Beilstein 01,468
Specific Gravity 1.12
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in benzene and toluene, practically insoluble in diethylether
Viscosity 44.9 mPa.s (25°C)
Formula Weight 194.23
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Tetraethylene glycol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.