missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium tetrafluoroborate, 99%

Product Code. 11419542
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
250 g
Unit Size:
100g
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419542 50 g 50g
11413950 5 g 5g
11423950 100 g 100g
11429542 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11419542 Supplier Thermo Scientific Alfa Aesar Supplier No. A10211.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 429-06-1 | C8H20BF4N | 217.06 g/mol

Tetraethylammonium tetrafluoroborate is used as a phase transfer catalyst in organic synthesis. It finds application as an electrolyte additive in the capillary electrophoretic separation of polyphenols found in grape seed extracts. It is also used to prepare other tetrabutylammonium salts in aqueous solutions. It is involved in the synthesis of conducting poly(thiophenes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium tetrafluoroborate is used as a phase transfer catalyst in organic synthesis. It finds application as an electrolyte additive in the capillary electrophoretic separation of polyphenols found in grape seed extracts. It is also used to prepare other tetrabutylammonium salts in aqueous solutions. It is involved in the synthesis of conducting poly(thiophenes).

Solubility
Soluble in water, alcohol and acetonitrile.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 429-06-1
Molecular Formula C8H20BF4N
Molecular Weight (g/mol) 217.06
MDL Number MFCD00011827
InChI Key XJRAKUDXACGCHA-UHFFFAOYSA-N
Synonym tetraethylammonium tetrafluoroborate, tetraethylazanium tetrafluoroborate, tetraethylammoniumtetrafluoroborate, acmc-1ago3, ksc238k1t, ethanaminium, n,n,n-triethyl-, tetrafluoroborate 1-, ethanaminium, n,n,n-triethyl-, tetrafluoroborate 1-1:1, tetraethyl ammonium tetrafluoroborate, tetraethylammonium tetra fluoro borate
PubChem CID 2724277
IUPAC Name tetraethylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC[N+](CC)(CC)CC

Specifications

Melting Point 370°C to 380°C (decomposition)
Linear Formula (CH3CH2)4NBF4
Quantity 50 g
UN Number UN2811
Beilstein 3917638
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol and acetonitrile.
Formula Weight 217.06
Percent Purity 99%
Chemical Name or Material Tetraethylammonium tetrafluoroborate
compliance-icons
Product Identifier
  • Tetraethylammonium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tetraethylammonium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.