missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium perchlorate, 98% (dry wt.), cont. ca 10% water

Product Code. p-7059146
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325138 10 g 10g
11335138 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325138 Supplier Thermo Scientific Alfa Aesar Supplier No. 044062.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2567-83-1 | C8H20ClNO4 | 229.70 g/mol

Tetraethylammonium perchlorate is used as supporting electrolyte in polarographic measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2567-83-1
Molecular Formula C8H20ClNO4
Molecular Weight (g/mol) 229.70
MDL Number MFCD00041977
InChI Key WGHUNMFFLAMBJD-UHFFFAOYSA-M
Synonym tetraethylammonium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate, tetraethylazanium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate 1:1, ammonium, tetraethyl-, perchlorate, tetraethylammoniumperchlorate, acmc-1ccq7, ksc203k4p, tetraethyl ammonium perchlorate, tetraethylammoniumperchlorate, dry
PubChem CID 2734932
IUPAC Name tetraethylazanium perchlorate
SMILES [O-][Cl](=O)(=O)=O.CC[N+](CC)(CC)CC

Specifications

Melting Point >300°C
Odor Odorless
Quantity 10 g
UN Number UN1479
Beilstein 3638093
Solubility Information Soluble in water,acetone,acetonitrile and dimethyl sulfoxide.
Formula Weight 229.71
Percent Purity 98%
Concentration or Composition (by Analyte or Components) Contains ≈ 10% water
Physical Form Wet Crystals
Assay (dry wt.)
Chemical Name or Material Tetraethylammonium perchlorate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetraethlammonium perchlorate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P362-Take off contaminated clothing.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.