missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium nitrate, 35% w/w aq. soln., Thermo Scientific™

Product Code. 11368948
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368948 250 mL 250mL
11378948 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368948 Supplier Thermo Scientific Alfa Aesar Supplier No. 041791.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1941-26-0
Molecular Formula C8H20N2O3
Molecular Weight (g/mol) 192.26
MDL Number MFCD00041978
InChI Key JTJKNAJRGLQKDZ-UHFFFAOYSA-N
Synonym tetraethylammonium nitrate, ethanaminium, n,n,n-triethyl-, nitrate, ammonium, tetraethyl-, nitrate, ethanaminium, n,n,n-triethyl-, nitrate 1:1, tetraethylazanium nitrate, tetraethyl ammonium nitrate, tetraethylammonium nitrate w/w aqueous solution
PubChem CID 74744
IUPAC Name tetraethylazanium;nitrate
SMILES [O-][N+]([O-])=O.CC[N+](CC)(CC)CC

Specifications

Density 1.029 g/mL
Boiling Point >100°C
Quantity 250 mL
UN Number UN3139
Beilstein 3918466
Formula Weight 192.26
Concentration or Composition (by Analyte or Components) 35% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetraethylammonium nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.