missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium hydroxide, 35% w/w aq. soln., Thermo Scientific Chemicals
39.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Tetraethylammonium hydroxide is a strong organic base and finds applications in photovoltaic chemicals, advanced ceramics, flat panel display chemicals, selective etching and transesterification catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Density | 1.023 g/mL |
Linear Formula | (CH3CH2)4NOH |
Refractive Index | 1.404 |
Quantity | 100 g |
UN Number | UN3267 |
Merck Index | 14,9201 |
Solubility Information | Miscible with water. |
Formula Weight | 147.26 |
Concentration or Composition (by Analyte or Components) | 35% w/w aq. soln. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'