missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hydroxide, 35% w/w aq. soln., Reagent Grade

Product Code. 11334668
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334668 500 g 500g
11314668 25 g 25g
11324668 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334668 Supplier Thermo Scientific Alfa Aesar Supplier No. 043023.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 77-98-5 | C8H21NO | 147.26 g/mol

Tetraethylammonium hydroxide is a strong organic base and finds applications in photovoltaic chemicals, advanced ceramics, flat panel display chemicals, selective etching and transesterification catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 77-98-5
Molecular Formula C8H21NO
Molecular Weight (g/mol) 147.26
MDL Number MFCD00009024
InChI Key LRGJRHZIDJQFCL-UHFFFAOYSA-M
Synonym tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
PubChem CID 6509
SMILES [OH-].CC[N+](CC)(CC)CC

Spezifikation

Density 1.023 g/mL
Boiling Point (decomposition)
Odor Amine-like
Linear Formula (CH3CH2)4NOH
Refractive Index 1.404
Quantity 500 g
UN Number UN3267
Beilstein 3912961
Sensitivity Air Sensitive
Merck Index 14,9201
Solubility Information Miscible with water.
IUPAC Name tetraethylazanium;hydroxide
Formula Weight 147.26
Grade Reagent
Concentration or Composition (by Analyte or Components) 35% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetraethylammonium hydroxide
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.