missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hydroxide, 25% w/w in methanol

Product Code. 11357178
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357178 10 g 10g
11347178 50 g 50g
11367178 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357178 Supplier Thermo Scientific Alfa Aesar Supplier No. 016328.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 77-98-5 | C8H21NO | 147.26 g/mol

It is used as a phase transfer catalyst, the template agent synthesized by molecular sieve and cleaning agents, as a surfactant, used to anisotropically etch silicon, in thermochemolysis, as a basic solvent in the development of acidic photo resist in the photolithography process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-98-5
Molecular Formula C8H21NO
Molecular Weight (g/mol) 147.26
MDL Number MFCD00009024
InChI Key LRGJRHZIDJQFCL-UHFFFAOYSA-M
Synonym tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
PubChem CID 6509
IUPAC Name tetraethylazanium;hydroxide
SMILES [OH-].CC[N+](CC)(CC)CC

Specifications

Density 0.88g/mL
Flash Point 16° (60°F)
Odor Like alcohol
Refractive Index 1.380
Quantity 10 g
UN Number UN2924
Beilstein 3912961
Sensitivity Air Sensitive
Merck Index 14,9201
Formula Weight 147.26
Concentration or Composition (by Analyte or Components) 25% w/w in methanol
Physical Form Liquid
Chemical Name or Material Tetraethylammonium hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.