Learn More
Tetraethylammonium hydroxide, 20 wt.% in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 176072500
Code : 29
Additional Details : CAS Number : 77-98-5 Weight : 0.25500kg Transport : UN number : 2922 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetraethylammonium hydroxide | |
77-98-5,7732-18-5 | |
1.0200g/mL | |
Plastic bottle | |
1.402 | |
MFCD00009024 | |
1.02 | |
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium | |
LRGJRHZIDJQFCL-UHFFFAOYSA-M | |
tetraethylazanium;hydroxide | |
6509 | |
≥20% |
20 wt.% in Water | |
Colorless to Yellow | |
102.0°C | |
C8H21NO | |
(C2H5)4NOH | |
250 mL | |
15, 9346 | |
Solubility in water: soluble. | |
[OH-].CC[N+](CC)(CC)CC | |
147.26 | |
147.26 | |
Liquid |
RUO – Research Use Only