missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hydroxide, 20% aq. soln.

Product Code. 11326736
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326736 500 g 500g
11316736 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326736 Supplier Thermo Scientific Alfa Aesar Supplier No. L03968.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 77-98-5 | C8H21NO | 147.26 g/mol

Tetraethylammonium hydroxide is used as structure Directing Agents (SDAs), in selective etching, photovoltaic chemicals, advanced ceramics, strong organic base, flat panel display chemicals and transesterification catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium hydroxide is used as structure Directing Agents (SDAs), in selective etching, photovoltaic chemicals, advanced ceramics, strong organic base, flat panel display chemicals and transesterification catalyst.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-98-5
Molecular Formula C8H21NO
Molecular Weight (g/mol) 147.26
MDL Number MFCD00009024
InChI Key LRGJRHZIDJQFCL-UHFFFAOYSA-M
Synonym tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
PubChem CID 6509
IUPAC Name tetraethylazanium;hydroxide
SMILES [OH-].CC[N+](CC)(CC)CC

Specifications

Density 1.023
Linear Formula (CH3CH2)4NOH
Refractive Index 1.373
Quantity 500 g
UN Number UN3267
Beilstein 3912961
Sensitivity Air Sensitive
Merck Index 14,9201
Solubility Information Fully miscible in water.
Formula Weight 147.26
Concentration or Composition (by Analyte or Components) 20% aq. soln.
Chemical Name or Material Tetraethylammonium hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.